siRNA Details
Gene Symbol: Eif1ax | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106963.1 | NP_001100433.1 | 2, 3 | 317 | ||
GenBank mRNA | BC168938.1 | - | 2, 3 | 190 | 7097069 | |
FQ217925.1 | - | 2, 3 | 282 | |||
FQ221854.1 | - | 2, 3 | 296 | |||
FQ222873.1 | - | 2, 3 | 315 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
