siRNA Details
Gene Symbol: RGD1561230 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106968.1 | NP_001100438.1 | 1 | 294 | ||
XM_006257206.2 | XP_006257268.1 | 2 | 421 | |||
XM_008773374.1 | XP_008771596.1 | 2 | 421 |
