siRNA Details
Gene Symbol: Zdhhc9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039016.2 | NP_001034105.2 | 9, 10 | 1273 | ||
XM_006257521.2 | XP_006257583.1 | 9, 10 | 1375 | |||
GenBank mRNA | AY886526.1 | - | 9, 10 | 1243 | ||
BC129070.1 | - | 9, 10 | 1273 | 7461054 |
