siRNA Details
Gene Symbol: Elf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191735.1 | NP_001178664.1 | 7 | 1138 | ||
XM_008773515.1 | XP_008771737.1 | 7 | 3573 | |||
XM_008773516.1 | XP_008771738.1 | 8 | 1406 | |||
XM_008773517.1 | XP_008771739.1 | 8 | 1604 | |||
XM_008773518.1 | XP_008771740.1 | 8 | 3333 |
