siRNA Details
Gene Symbol: MGC105649 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008518.2 | NP_001008518.1 | 3, 4 | 171 | ||
XM_008762164.1 | XP_008760386.1 | 2, 3 | 262 | |||
GenBank mRNA | BC086337.1 | - | 2, 3 | 147 | 7301704 | |
CB585387.1 | - | 3, 4 | 176 | |||
FQ216036.1 | - | 2, 3 | 268 | |||
FQ223264.1 | - | 3, 4 | 167 |

More Information
Other Species Targeted

