siRNA Details
Gene Symbol: Eef2kmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106975.1 | NP_001100445.1 | 6 | 574 | ||
XM_006245785.2 | XP_006245847.1 | 5 | 540 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
