siRNA Details
Gene Symbol: Srrm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277154.1 | NP_001264083.1 | 12 | 6521 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Srrm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277154.1 | NP_001264083.1 | 12 | 6521 |