siRNA Details
Gene Symbol: Noxo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106986.1 | NP_001100456.1 | 6 | 1034 | ||
XM_006245948.1 | XP_006246010.1 | 6 | 659 | |||
XM_006245949.1 | XP_006246011.1 | 5 | 1046 | |||
XM_006245950.2 | XP_006246012.1 | 5 | 1046 | |||
XM_008767566.1 | XP_008765788.1 | 5 | 1046 |
