siRNA Details
Gene Symbol: Mapk8ip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100673.1 | NP_001094143.1 | 13 | 1554 | ||
XM_006245953.2 | XP_006246015.1 | 15 | 1732 | |||
XM_006245954.2 | XP_006246016.1 | 15 | 1729 | |||
XM_006245955.2 | XP_006246017.1 | 14 | 1707 | |||
XM_006245956.2 | XP_006246018.1 | 14 | 1705 | |||
XM_006245957.2 | XP_006246019.1 | 15 | 1705 | |||
XM_006245958.2 | XP_006246020.1 | 13 | 1686 | |||
XM_006245959.2 | XP_006246021.1 | 15 | 1639 | |||
XM_006245960.2 | XP_006246022.1 | 15 | 1636 | |||
XM_006245961.2 | XP_006246023.1 | 14 | 1614 | |||
XM_006245962.2 | XP_006246024.1 | 14 | 1611 | |||
XM_006245963.2 | XP_006246025.1 | 13 | 1596 | |||
XM_008767567.1 | XP_008765789.1 | 14 | 1713 | |||
XM_008767568.1 | XP_008765790.1 | 10 | 1404 | |||
XM_008767569.1 | XP_008765791.1 | 10 | 941 | |||
GenBank mRNA | DQ377222.1 | - | 13 | 1554 |
