siRNA Details
Gene Symbol: Haghl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750790.3 | XP_003750838.1 | 8 | 852 | ||
XM_006246064.2 | XP_006246126.1 | 7 | 933 | |||
XM_006246065.1 | XP_006246127.1 | 4 | 543 | |||
XM_008767617.1 | XP_008765839.1 | 5 | 941 | |||
GenBank mRNA | BC076383.1 | - | 5 | 980 | ||
BC161824.1 | - | 4 | 1179 |

More Information
Other Species Targeted

Gene Ontology Categories:
