siRNA Details
Gene Symbol: Fbxw11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106993.1 | NP_001100463.1 | 5 | 582 | ||
XM_006246092.2 | XP_006246154.1 | 5 | 588 | |||
XM_006246093.2 | XP_006246155.1 | 5 | 579 | |||
XM_006246094.2 | XP_006246156.1 | 4 | 516 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001271347, NM_001271349, NM_134015, NM_001271348, AK168667, and others.
Gene Ontology Categories:
