siRNA Details
Gene Symbol: Slu7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100550.1 | NP_001094020.1 | 16 | 1734 | ||
XM_008767640.1 | XP_008765862.1 | 15 | 2509 | |||
XM_008767641.1 | XP_008765863.1 | 16 | 2181 | |||
XM_008767642.1 | XP_008765864.1 | 16 | 2386 | |||
GenBank mRNA | BC158865.1 | - | 16 | 1700 | 7938574 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
