siRNA Details
Gene Symbol: Nipal4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106995.1 | NP_001100465.1 | 5 | 552 | ||
GenBank mRNA | FQ211728.1 | - | 5 | 621 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Nipal4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106995.1 | NP_001100465.1 | 5 | 552 | ||
GenBank mRNA | FQ211728.1 | - | 5 | 621 |