siRNA Details
Gene Symbol: RGD1310352 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106999.1 | NP_001100469.1 | 3 | 755 | ||
XM_006246261.2 | XP_006246323.1 | 3 | 767 | |||
GenBank mRNA | BC079420.1 | - | 3 | 724 |

More Information
Other Species Targeted

