siRNA Details
Gene Symbol: Rapgef6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107003.1 | NP_001100473.1 | 6 | 584 | ||
XM_006246277.2 | XP_006246339.1 | 6 | 633 | |||
XM_006246278.2 | XP_006246340.1 | 6 | 632 | |||
XM_006246279.2 | XP_006246341.1 | 6 | 632 | |||
XM_006246280.2 | XP_006246342.1 | 6 | 632 |

More Information
Other Species Targeted

Gene Ontology Categories:
