siRNA Details
Gene Symbol: Guk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013115.2 | NP_001013133.2 | 2, 3 | 172 | ||
XM_006246477.2 | XP_006246539.1 | 3, 4 | 309 | |||
XM_006246478.2 | XP_006246540.1 | 2, 3 | 274 | |||
XM_006246479.2 | XP_006246541.1 | 1, 2 | 117 | |||
XM_006246483.1 | XP_006246545.1 | 3, 4 | 224 | |||
XM_008767812.1 | XP_008766034.1 | 3, 4 | 358 | |||
XM_008767813.1 | XP_008766035.1 | 2, 3 | 933 | |||
XM_008767814.1 | XP_008766036.1 | 2, 3 | 182 | |||
XM_008767815.1 | XP_008766037.1 | 2, 3 | 929 | |||
GenBank mRNA | AF354443.1 | - | 2, 3 | 123 | ||
FM065046.1 | - | 2, 3 | 172 |
