siRNA Details
Gene Symbol: RGD1304587 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100551.1 | NP_001094021.1 | 1 | 250 | ||
XM_006246486.2 | XP_006246548.1 | 1 | 1339 | |||
GenBank mRNA | BC086951.1 | - | 1 | 186 | ||
BC162001.1 | - | 1 | 250 | 9088363 |
