siRNA Details
Gene Symbol: Tmem11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164543.1 | NP_001158015.1 | 2 | 1130 | ||
NM_001166165.1 | NP_001159637.1 | 3 | 1417 | |||
GenBank mRNA | BC158725.1 | - | 2 | 1130 | 9090994 | |
FQ220528.1 | - | 2 | 894 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
