siRNA Details
Gene Symbol: Ctc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107010.1 | NP_001100480.1 | 17 | 3048 | ||
XM_006246699.2 | XP_006246761.1 | 17 | 3094 | |||
XM_006246700.1 | XP_006246762.1 | 14 | 2274 |

More Information
Other Species Targeted

Gene Ontology Categories:
