siRNA Details
Gene Symbol: Dnah2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008767898.1 | XP_008766120.1 | 49 | 7912 | ||
XM_008767899.1 | XP_008766121.1 | 49 | 7748 | |||
XM_008767900.1 | XP_008766122.1 | 43 | 6711 | |||
GenBank mRNA | AY903242.1 | - | 38 | 5811 | ||
AY903243.1 | - | 38 | 5811 | |||
AY903244.1 | - | 38 | 5811 |
