siRNA Details
Gene Symbol: Dnah2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008767898.1 | XP_008766120.1 | 29, 30 | 5072 | ||
XM_008767899.1 | XP_008766121.1 | 29, 30 | 4908 | |||
XM_008767900.1 | XP_008766122.1 | 23, 24 | 3871 | |||
GenBank mRNA | AY903242.1 | - | 18, 19 | 2971 | ||
AY903243.1 | - | 18, 19 | 2971 | |||
AY903244.1 | - | 18, 19 | 2971 |
