siRNA Details
Gene Symbol: Spag7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107016.1 | NP_001100486.1 | 3, 4 | 375 | ||
GenBank mRNA | FQ233279.1 | - | 3, 4 | 247 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
