siRNA Details
Gene Symbol: Fbxo39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039018.1 | NP_001034107.1 | 2 | 1115 | ||
XM_006246727.2 | XP_006246789.1 | 3 | 1372 | |||
XM_006246729.2 | XP_006246791.1 | 3 | 1212 | |||
GenBank mRNA | BC082089.1 | - | 3 | 1165 | ||
BC100079.1 | - | 2 | 1115 | 7132945 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
