siRNA Details
Gene Symbol: Cyb5d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007671.1 | NP_001007672.1 | 2 | 512 | ||
XM_006246730.2 | XP_006246792.1 | 2 | 290 | |||
GenBank mRNA | BC079177.1 | - | 2 | 512 | 7130324 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
