siRNA Details
Gene Symbol: Tmem199 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024992.1 | NP_001020163.1 | 1 | 198 | ||
GenBank mRNA | BC091246.1 | - | 1 | 198 | 7366299 | |
FQ215691.1 | - | 1 | 202 | |||
FQ216010.1 | - | 1 | 222 | |||
FQ223430.1 | - | 1 | 222 |

More Information
Other Species Targeted

