siRNA Details
Gene Symbol: Atad5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006246963.2 | XP_006247025.1 | 7 | 2989 | ||
XM_220750.8 | XP_220750.6 | 7 | 2989 |

More Information
Other Species Targeted

Gene Ontology Categories:
