siRNA Details
Gene Symbol: Lig3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012011.1 | NP_001012011.1 | 13, 14 | 2075 | ||
XM_006246989.2 | XP_006247051.1 | 13, 14 | 2109 | |||
XM_006246990.2 | XP_006247052.1 | 13, 14 | 2095 | |||
XM_008768020.1 | XP_008766242.1 | 12, 13 | 2978 | |||
XM_008768021.1 | XP_008766243.1 | 12, 13 | 2981 | |||
XM_008768022.1 | XP_008766244.1 | 12, 13 | 2981 | |||
XM_008768023.1 | XP_008766245.1 | 13, 14 | 2109 | |||
GenBank mRNA | BC085959.1 | - | 13, 14 | 2075 | 7135421 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_010716, BC049240, AK156825, U66057, AK156030, and others.
Gene Ontology Categories:
