siRNA Details
Gene Symbol: Usp32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107032.1 | NP_001100502.1 | 10 | 1195 | ||
XM_006247034.1 | XP_006247096.1 | 15 | 1608 | |||
XM_008768035.1 | XP_008766257.1 | 15 | 1608 |

More Information
Other Species Targeted

Gene Ontology Categories:
