siRNA Details
Gene Symbol: Tbx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107034.1 | NP_001100504.1 | 4 | 473 | ||
XM_006247036.2 | XP_006247098.1 | 4 | 571 | |||
XM_006247037.2 | XP_006247099.1 | 2 | 2493 | |||
XM_008768036.1 | XP_008766258.1 | 4 | 360 | |||
XM_008768037.1 | XP_008766259.1 | 1 | 1172 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_011536, NM_172798, U57329, AK028407, AK139949, and others.
Gene Ontology Categories:
