siRNA Details
Gene Symbol: Rnf43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135921.1 | NP_001129393.1 | 7 | 884 | ||
XM_006247090.2 | XP_006247152.1 | 8 | 2369 | |||
XM_006247091.2 | XP_006247153.1 | 8 | 2144 | |||
XM_006247092.2 | XP_006247154.1 | 7 | 2889 | |||
XM_006247094.2 | XP_006247156.1 | 6 | 2245 | |||
XM_008768038.1 | XP_008766260.1 | 7 | 3280 | |||
XM_008768039.1 | XP_008766261.1 | 5 | 2923 | |||
XM_008768040.1 | XP_008766262.1 | 6 | 2825 | |||
XM_008768041.1 | XP_008766263.1 | 6 | 2473 |
