siRNA Details
Gene Symbol: Mpo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006247095.2 | XP_006247157.1 | 9 | 1053 | ||
XM_006247096.2 | XP_006247158.1 | 9 | 1055 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
