siRNA Details
Gene Symbol: Epx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107037.1 | NP_001100507.1 | 10 | 1797 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Epx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107037.1 | NP_001100507.1 | 10 | 1797 |