siRNA Details
Gene Symbol: Sgca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107039.1 | NP_001100509.1 | 4 | 532 | ||
XM_006247191.2 | XP_006247253.1 | 3 | 356 | |||
XM_006247192.1 | XP_006247254.1 | 3 | 260 | |||
GenBank mRNA | FQ215532.1 | - | 3 | 318 |
