siRNA Details
Gene Symbol: Abi3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013118.1 | NP_001013136.1 | 8 | 1264 | ||
XM_008768045.1 | XP_008766267.1 | 8 | 1006 | |||
XM_008768046.1 | XP_008766268.1 | 8 | 1003 | |||
GenBank mRNA | BC079102.1 | - | 8 | 1264 | 7124908 | |
FQ225352.1 | - | 8 | 995 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
