siRNA Details
Gene Symbol: Ube2z | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037643.2 | NP_001032732.2 | 7 | 1006 | ||
GenBank mRNA | BC107663.1 | - | 7 | 1006 | 7936709 |

More Information
Other Species Targeted

Gene Ontology Categories:
