siRNA Details
Gene Symbol: Ikzf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107047.1 | NP_001100517.1 | 4 | 316 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ikzf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107047.1 | NP_001100517.1 | 4 | 316 |