siRNA Details
Gene Symbol: Rapgefl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008768202.1 | XP_008766424.1 | 3, 4 | 1121 | ||
XM_008768203.1 | XP_008766425.1 | 3, 4 | 1122 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
