siRNA Details
Gene Symbol: Dnajc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_213625.1 | NP_998790.1 | 4 | 373 | ||
XM_006247311.2 | XP_006247373.1 | 5 | 734 | |||
XM_006247312.2 | XP_006247374.1 | 4 | 454 | |||
XM_006247313.1 | XP_006247375.1 | 3 | 294 | |||
GenBank mRNA | AB178470.1 | - | 4 | 373 | ||
BC093600.1 | - | 3 | 289 | 7316009 | ||
FQ212948.1 | - | 4 | 404 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_019795, AK145506, BC055729, AY323828, AK145739, and others.
Gene Ontology Categories:
