siRNA Details
Gene Symbol: Zfp385c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006247466.2 | XP_006247528.1 | 6 | 2473 | ||
XM_008768216.1 | XP_008766438.1 | 7 | 1196 | |||
XM_008768217.1 | XP_008766439.1 | 6 | 2473 | |||
XM_008768218.1 | XP_008766440.1 | 6 | 1013 | |||
GenBank mRNA | BC162044.1 | - | 5 | 921 |
