siRNA Details
Gene Symbol: Ezh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107051.1 | NP_001100521.1 | 11, 12 | 1483 | ||
XM_006247318.2 | XP_006247380.1 | 12, 13 | 1529 | |||
XM_006247319.2 | XP_006247381.1 | 12, 13 | 1593 | |||
XM_008768052.1 | XP_008766274.1 | 12, 13 | 1534 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_007970, U60453, AK138942, AK140694, AF483490, and others.
Gene Ontology Categories:
