siRNA Details
Gene Symbol: Ezh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107051.1 | NP_001100521.1 | 18 | 2110 | ||
XM_006247318.2 | XP_006247380.1 | 19 | 2156 | |||
XM_006247319.2 | XP_006247381.1 | 19 | 2220 | |||
XM_008768052.1 | XP_008766274.1 | 19 | 2161 |

More Information
Other Species Targeted

Gene Ontology Categories:
