siRNA Details
Gene Symbol: Nbr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024765.1 | NP_001019936.1 | 6, 7 | 392 | ||
XM_006247321.2 | XP_006247383.1 | 6, 7 | 674 | |||
XM_006247322.2 | XP_006247384.1 | 6, 7 | 589 | |||
XM_006247323.2 | XP_006247385.1 | 6, 7 | 446 | |||
XM_006247324.2 | XP_006247386.1 | 6, 7 | 393 | |||
XM_006247325.2 | XP_006247387.1 | 6, 7 | 549 | |||
XM_006247326.2 | XP_006247388.1 | 6, 7 | 544 | |||
GenBank mRNA | BC095903.1 | - | 6, 7 | 392 | 7096697 | |
FQ211883.1 | - | 6, 7 | 392 |

More Information
Other Species Targeted

Gene Ontology Categories:
