siRNA Details
Gene Symbol: Nags | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107053.1 | NP_001100523.1 | 7 | 1593 | ||
XM_006247328.2 | XP_006247390.1 | 8 | 2413 | |||
XM_008768055.1 | XP_008766277.1 | 8 | 2413 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
