siRNA Details
Gene Symbol: Tmem101 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191650.1 | NP_001178579.1 | 2 | 203 | ||
GenBank mRNA | BC086567.1 | - | 2 | 151 | ||
DY310329.1 | - | 2 | 203 |

More Information
Other Species Targeted

Gene Ontology Categories:
