siRNA Details
Gene Symbol: Rundc3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198758.2 | NP_942053.2 | 11 | 2003 | ||
XM_006247335.2 | XP_006247397.1 | 11 | 2001 | |||
XM_006247337.2 | XP_006247399.1 | 11 | 1962 | |||
XM_008768056.1 | XP_008766278.1 | 11 | 1989 | |||
XM_008768058.1 | XP_008766280.1 | 11 | 1986 | |||
GenBank mRNA | AF288611.1 | - | 5 | 927 | ||
BC127472.1 | - | 10 | 1587 | |||
BC158572.1 | - | 8 | 1376 |
