siRNA Details
Gene Symbol: March10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013973.1 | NP_001013995.1 | 5 | 482 | ||
XM_006247579.2 | XP_006247641.1 | 5 | 650 | |||
XM_006247580.2 | XP_006247642.1 | 6 | 769 | |||
XM_006247585.2 | XP_006247647.1 | 5 | 627 | |||
GenBank mRNA | AB552845.1 | - | 3 | 227 | ||
AB552846.1 | - | 3 | 227 | |||
BC083567.1 | - | 5 | 482 | 7112851 |
