siRNA Details
Gene Symbol: Tanc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191653.1 | NP_001178582.1 | 8, 9 | 1214 | ||
XM_008768351.1 | XP_008766573.1 | 9, 10 | 1333 | |||
XM_008768352.1 | XP_008766574.1 | 9, 10 | 1323 | |||
XM_008768353.1 | XP_008766575.1 | 9, 10 | 1333 | |||
XM_008768354.1 | XP_008766576.1 | 8, 9 | 1222 | |||
XM_008768355.1 | XP_008766577.1 | 9, 10 | 1222 | |||
XM_008768356.1 | XP_008766578.1 | 8, 9 | 1111 |
