siRNA Details
Gene Symbol: Arsg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047877.1 | NP_001041342.1 | 1 | 70 | ||
XM_006247599.2 | XP_006247661.1 | 3 | 2270 | |||
XM_006247600.2 | XP_006247662.1 | 2 | 2127 | |||
XM_006247601.2 | XP_006247663.1 | 4 | 2496 | |||
XM_006247602.2 | XP_006247664.1 | 3 | 1058 | |||
XM_006247603.2 | XP_006247665.1 | 3 | 912 | |||
XM_006247604.2 | XP_006247666.1 | 3 | 2328 | |||
XM_006247606.2 | XP_006247668.1 | 2 | 2185 | |||
XM_006247607.2 | XP_006247669.1 | 2 | 1097 | |||
XM_006247608.2 | XP_006247670.1 | 1 | 747 | |||
XM_008768368.1 | XP_008766590.1 | 4 | 2439 | |||
XM_008768369.1 | XP_008766591.1 | 5 | 1903 |
