siRNA Details
Gene Symbol: Gna13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013119.1 | NP_001013137.1 | 4 | 853 | ||
GenBank mRNA | AY553631.1 | - | 4 | 853 | ||
AY553632.1 | - | 4 | 853 | |||
DQ120481.1 | - | 4 | 853 | |||
DQ120482.1 | - | 4 | 853 |
