siRNA Details
Gene Symbol: Nat9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134835.1 | NP_001128307.1 | 1 | 448 | ||
XM_006247714.2 | XP_006247776.1 | 2 | 307 | |||
GenBank mRNA | BC158710.1 | - | 1 | 448 | 9035021 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
